Order Entry
Italy
ContactUsLinkComponent
Ethyl 8-chloro-4-hydroxy-6-methylquinoline-3-carboxylate ≥98%
Ethyl 8-chloro-4-hydroxy-6-methylquinoline-3-carboxylate ≥98%
Catalog # APOSOR183505-1G
Supplier:  Apollo Scientific
CAS Number:  
Ethyl 8-chloro-4-hydroxy-6-methylquinoline-3-carboxylate ≥98%
Catalog # APOSOR183505-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :265.692 g/mol
Storage Temperature:Refrigerator
CAS Number:338795-13-4
Formula:C₁₃H₁₂ClNO₃
MDL Number:MFCD00173392