Order Entry
Italy
ContactUsLinkComponent
6-(1H-Pyrazol-3-yl)-4H-1,4-benzoxazin-3-one
6-(1H-Pyrazol-3-yl)-4H-1,4-benzoxazin-3-one
Catalog # APOSOR183463-1G
Supplier:  Apollo Scientific
CAS Number:  
6-(1H-Pyrazol-3-yl)-4H-1,4-benzoxazin-3-one
Catalog # APOSOR183463-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :215.208 g/mol
Storage Temperature:Ambient
CAS Number:948551-31-3
Formula:C₁₁H₉N₃O₂
MDL Number:MFCD05664896