Order Entry
Italy
ContactUsLinkComponent
2-Methyl-3-(1H-pyrazol-3-yl)imidazo[1,2-a]pyridine
2-Methyl-3-(1H-pyrazol-3-yl)imidazo[1,2-a]pyridine
Codice catalogo APOSOR183431-1G
Supplier:  Apollo Scientific
CAS Number:  
2-Methyl-3-(1H-pyrazol-3-yl)imidazo[1,2-a]pyridine
Codice catalogo APOSOR183431-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :198.224 g/mol
Storage Temperature:Ambient
CAS Number:264226-27-9
Formula:C₁₁H₁₀N₄
MDL Number:MFCD00269358