Order Entry
Italy
ContactUsLinkComponent
3-Chlorophenylglyoxal hydrate 95%
Irritant
3-Chlorophenylglyoxal hydrate 95%
Catalog # APOSOR1625-250MG
Supplier:  Apollo Scientific
CAS Number:  
Irritant
3-Chlorophenylglyoxal hydrate 95%
Catalog # APOSOR1625-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :186.592 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:177288-16-3
Formula:C₈H₅ClO₂·H₂O
MDL Number:MFCD08705874