
1,3-Thiazole-4-thiocarboxamide
Catalog # APOSOR15911-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:144.218 g/mol
:GHS07
Storage Temperature:Refrigerator
CAS Number:80653-66-3
Formula:C₄H₄N₂S₂
MDL Number:MFCD04115283