3-[(Pyrazin-2-yl)oxy]benzaldehyde
Bottle, 100 mg
Codice catalogo APOSOR12897-100MG
Nome del fornitore: Apollo Scientific
3-[(Pyrazin-2-yl)oxy]benzaldehyde
Codice catalogo APOSOR12897-100MG
Nome del fornitore: Apollo Scientific
Numero CAS: 887344-44-7
Jump to:
Product Details & Documents
Synonyms: 2-(3-Formylphenoxy)-1,4-diazine, 2-(3-Formylphenoxy)pyrazine
- CAS Number:887344-44-7
- Formula:C₁₁H₈N₂O₂
- MDL Number:MFCD03840111
- Molecular Weight:200.193 g/mol
- Storage Temperature:Ambient
Specifications
Specifications
- Pack type:Bottle
- Pk:100 mg
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



