Ethyl (2-acetyl-4-chlorophenoxy)acetate
Catalog # APOSOR111599-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:256.682 g/mol
Storage Temperature:Ambient
CAS Number:100119-57-1
Formula:C₁₂H₁₃ClO₄
MDL Number:MFCD08059210