Order Entry
Italy
ContactUsLinkComponent
Ethyl (2-acetyl-5-bromophenoxy)acetate
Ethyl (2-acetyl-5-bromophenoxy)acetate
Catalog # APOSOR111597-1G
Supplier:  Apollo Scientific
CAS Number:  
Ethyl (2-acetyl-5-bromophenoxy)acetate
Catalog # APOSOR111597-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :301.133 g/mol
Storage Temperature:Ambient
CAS Number:91721-00-5
Formula:C₁₂H₁₃BrO₄
MDL Number:MFCD27932676