Order Entry
Italy
ContactUsLinkComponent
2,2'-Bis(3-hydroxy-1,4-naphthoquinone)
Irritant
2,2'-Bis(3-hydroxy-1,4-naphthoquinone)
Catalog # APOSOR11131-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
2,2'-Bis(3-hydroxy-1,4-naphthoquinone)
Catalog # APOSOR11131-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
Melting Pt:249…251 °C
 :346.29 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:33440-64-1
Formula:C₂₀H₁₀O₆
MDL Number:MFCD00191788