Order Entry
Italy
ContactUsLinkComponent
 
N-(4-Chloro-2,5-dimethoxyphenyl)azepane-1-carbothioamide
Catalog # APOSOR111239-1G
Supplier:  Apollo Scientific
CAS Number:  
N-(4-Chloro-2,5-dimethoxyphenyl)azepane-1-carbothioamide
Catalog # APOSOR111239-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :328.857 g/mol
Storage Temperature:Ambient
Formula:C₁₅H₂₁ClN₂O₂S
MDL Number:MFCD30188165