(2-Chlorophenyl)(1-oxo-1,3-dihydro-2H-isoindol-2-yl)acetic acid
Catalog # APOSOR111232-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:301.724 g/mol
Storage Temperature:Ambient
Formula:C₁₆H₁₂ClNO₃
MDL Number:MFCD30188160