1-(1,3-Benzodioxol-5-yl)imidazolidin-2-one
Catalog # APOSOR111226-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:206.198 g/mol
Storage Temperature:Ambient
Formula:C₁₀H₁₀N₂O₃
MDL Number:MFCD23696661