
tert-Butyl 4-(cyanoacetyl)piperazine-1-carboxylate
Catalog # APOSOR111203-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:253.298 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:159873-21-9
Formula:C₁₂H₁₉N₃O₃
MDL Number:MFCD20731084