N,N,1,2-Tetramethyl-1H-imidazole-4-sulphonamide
Catalog # APOSOR111186-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:203.262 g/mol
Storage Temperature:Ambient
CAS Number:936083-53-3
Formula:C₇H₁₃N₃O₂S
MDL Number:MFCD09965509