5-Chloro-3-(morpholin-4-ylsulphonyl)pyridin-2-amine
Catalog # APOSOR111127-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:277.728 g/mol
Storage Temperature:Ambient
CAS Number:1858255-77-2
Formula:C₉H₁₂ClN₃O₃S
MDL Number:MFCD28954648