Order Entry
Italy
ContactUsLinkComponent
4-(2,5-Dimethyl-4-nitro-pyrazol-3-yl)morpholine
4-(2,5-Dimethyl-4-nitro-pyrazol-3-yl)morpholine
Catalog # APOSOR111099-1G
Supplier:  Apollo Scientific
CAS Number:  
4-(2,5-Dimethyl-4-nitro-pyrazol-3-yl)morpholine
Catalog # APOSOR111099-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :226.232 g/mol
Storage Temperature:Ambient
CAS Number:312311-18-5
Formula:C₉H₁₄N₄O₃
MDL Number:MFCD00121959