Order Entry
Italy
ContactUsLinkComponent
Ethyl 1-(4-methoxybenzyl)piperidine-4-carboxylate
Ethyl 1-(4-methoxybenzyl)piperidine-4-carboxylate
Catalog # APOSOR111077-1G
Supplier:  Apollo Scientific
CAS Number:  
Ethyl 1-(4-methoxybenzyl)piperidine-4-carboxylate
Catalog # APOSOR111077-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :277.359 g/mol
Storage Temperature:Ambient
CAS Number:148729-11-7
Formula:C₁₆H₂₃NO₃
MDL Number:MFCD00811156