Order Entry
Italy
ContactUsLinkComponent
3-[(2,6-Dimethylmorpholin-4-yl)methyl]benzonitrile
3-[(2,6-Dimethylmorpholin-4-yl)methyl]benzonitrile
Catalog # APOSOR111038-1G
Supplier:  Apollo Scientific
CAS Number:  
3-[(2,6-Dimethylmorpholin-4-yl)methyl]benzonitrile
Catalog # APOSOR111038-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :230.306 g/mol
Storage Temperature:Ambient
CAS Number:1016709-98-0
Formula:C₁₄H₁₈N₂O
MDL Number:MFCD09928897