Order Entry
Italy
ContactUsLinkComponent
 
Ethyl 3-morpholin-4-ylquinoxaline-2-carboxylate
Catalog # APOSOR110928-1G
Supplier:  Apollo Scientific
CAS Number:  
Ethyl 3-morpholin-4-ylquinoxaline-2-carboxylate
Catalog # APOSOR110928-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :287.314 g/mol
Storage Temperature:Ambient
CAS Number:108222-18-0
Formula:C₁₅H₁₇N₃O₃
MDL Number:MFCD27995620