Order Entry
Italy
ContactUsLinkComponent
1,4,6,7-Tetrahydropyrano[4,3-c]pyrazole
Irritant
1,4,6,7-Tetrahydropyrano[4,3-c]pyrazole
Catalog # APOSOR110924-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
1,4,6,7-Tetrahydropyrano[4,3-c]pyrazole
Catalog # APOSOR110924-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :124.141 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:258353-57-0
Formula:C₆H₈N₂O
MDL Number:MFCD18250670