Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Italy
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
6-tert-Butyl-3H-thieno[3,2-d]pyrimidin-4-one
6-tert-Butyl-3H-thieno[3,2-d]pyrimidin-4-one
6-tert-Butyl-3H-thieno[3,2-d]pyrimidin-4-one
6-tert-Butyl-3H-thieno[3,2-d]pyrimidin-4-one
APOSOR110921-1G
:
Apollo Scientific
:
6-tert-Butyl-3H-thieno[3,2-d]pyrimidin-4-one
APOSOR110921-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
208.28 g/mol
:
Ambient
:
215928-65-7
:
C₁₀H₁₂N₂OS
:
MFCD01570181