Order Entry
Italy
ContactUsLinkComponent
Ethyl 3-bromoimidazo[1,2-a]pyridine-7-carboxylate
Irritant
Ethyl 3-bromoimidazo[1,2-a]pyridine-7-carboxylate
Catalog # APOSOR110864-250MG
Supplier:  Apollo Scientific
CAS Number:  
Irritant
Ethyl 3-bromoimidazo[1,2-a]pyridine-7-carboxylate
Catalog # APOSOR110864-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :269.095 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:1251033-23-4
Formula:C₁₀H₉BrN₂O₂
MDL Number:MFCD22551971