
3-Amino-3-(1,3-benzodioxol-5-yl)propan-1-ol
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:113511-45-8
:
Formula:C₁₀H₁₃NO₃
MDL Number:MFCD03412636
:195.215 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together