Order Entry
Italy
ContactUsLinkComponent
Methyl 4-amino-2-(4-methylpiperazin-1-yl)-1,3-thiazole-5-carboxylate
Methyl 4-amino-2-(4-methylpiperazin-1-yl)-1,3-thiazole-5-carboxylate
Catalog # APOSOR110764-1G
Supplier:  Apollo Scientific
CAS Number:  
Methyl 4-amino-2-(4-methylpiperazin-1-yl)-1,3-thiazole-5-carboxylate
Catalog # APOSOR110764-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :256.325 g/mol
Storage Temperature:Ambient
Formula:C₁₀H₁₆N₄O₂S
MDL Number:MFCD18804168