Order Entry
Italy
ContactUsLinkComponent
4-[(4-Methyl-1,3-thiazol-5-yl)sulphonyl]morpholine
4-[(4-Methyl-1,3-thiazol-5-yl)sulphonyl]morpholine
Catalog # APOSOR110677-1G
Supplier:  Apollo Scientific
CAS Number:  
4-[(4-Methyl-1,3-thiazol-5-yl)sulphonyl]morpholine
Catalog # APOSOR110677-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :248.322 g/mol
Storage Temperature:Ambient
CAS Number:1427460-75-0
Formula:C₈H₁₂N₂O₃S₂
MDL Number:MFCD21602389