Order Entry
Italy
ContactUsLinkComponent
N,N-Diethyl-N'-(4-morpholin-4-ylphenyl)thiourea
N,N-Diethyl-N'-(4-morpholin-4-ylphenyl)thiourea
Catalog # APOSOR110663-1G
Supplier:  Apollo Scientific
CAS Number:  
N,N-Diethyl-N'-(4-morpholin-4-ylphenyl)thiourea
Catalog # APOSOR110663-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :293.428 g/mol
Storage Temperature:Ambient
CAS Number:1427460-42-1
Formula:C₁₅H₂₃N₃OS
MDL Number:MFCD20037420