Order Entry
Italy
ContactUsLinkComponent
Methyl 3-amino-4-({2-[(tert-butoxycarbonyl)amino]ethyl}amino)benzoate
Methyl 3-amino-4-({2-[(tert-butoxycarbonyl)amino]ethyl}amino)benzoate
Catalog # APOSOR110655-1G
Supplier:  Apollo Scientific
CAS Number:  
Methyl 3-amino-4-({2-[(tert-butoxycarbonyl)amino]ethyl}amino)benzoate
Catalog # APOSOR110655-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :309.361 g/mol
Storage Temperature:Ambient
CAS Number:1198093-71-8
Formula:C₁₅H₂₃N₃O₄
MDL Number:MFCD19442760