Order Entry
Italy
ContactUsLinkComponent
4-(4-Morpholin-4-ylphenyl)-1,3-thiazol-2-amine
Irritant
4-(4-Morpholin-4-ylphenyl)-1,3-thiazol-2-amine
Catalog # APOSOR110631-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
4-(4-Morpholin-4-ylphenyl)-1,3-thiazol-2-amine
Catalog # APOSOR110631-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :261.343 g/mol
 :GHS07
Storage Temperature:Refrigerator
CAS Number:436152-07-7
Formula:C₁₃H₁₅N₃OS
MDL Number:MFCD17168293