
tert-Butyl 4-[2-(2-bromophenoxy)ethyl]piperazine-1-carboxylate
Catalog # APOSOR110585-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:385.296 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:1227954-89-3
Formula:C₁₇H₂₅BrN₂O₃
MDL Number:MFCD11973602