Ethyl 1-methyl-1H-indole-4-carboxylate
Catalog # APOSOR110448-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:203.237 g/mol
Storage Temperature:Ambient
Formula:C₁₂H₁₃NO₂
MDL Number:MFCD20037415