
N-(1-Methyl-1H-indazol-7-yl)thiourea
Catalog # APOSOR110401-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:206.268 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:41968-38-1
Formula:C₉H₁₀N₄S
MDL Number:MFCD17168291