Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Italy
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-Methyl-7-nitro-2H-indazole
2-Methyl-7-nitro-2H-indazole
2-Methyl-7-nitro-2H-indazole
APOSOR110332-1G
:
Apollo Scientific
:
2-Methyl-7-nitro-2H-indazole
APOSOR110332-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
177.16 g/mol
:
Ambient
:
13436-58-3
:
C₈H₇N₃O₂
:
MFCD01719664