Methyl 3-(1H-pyrrol-1-yl)-1-benzothiophene-2-carboxylate
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:80066-96-2
:
Formula:C₁₄H₁₁NO₂S
MDL Number:MFCD02083299
:257.308 g/mol
Storage Temperature:Ambient
Frequently Bought Together