Order Entry
Italy
ContactUsLinkComponent
Ethyl 6-bromo-3-(1H-pyrrol-1-yl)-1-benzothiophene-2-carboxylate
Ethyl 6-bromo-3-(1H-pyrrol-1-yl)-1-benzothiophene-2-carboxylate
Catalog # APOSOR110215-1G
Supplier:  Apollo Scientific
CAS Number:  
Ethyl 6-bromo-3-(1H-pyrrol-1-yl)-1-benzothiophene-2-carboxylate
Catalog # APOSOR110215-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :350.23 g/mol
Storage Temperature:Ambient
Formula:C₁₅H₁₂BrNO₂S
MDL Number:MFCD11615814