Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Italy
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
1-Methyl-5-nitro-1H-indazol-3-amine
1-Methyl-5-nitro-1H-indazol-3-amine
1-Methyl-5-nitro-1H-indazol-3-amine
APOSOR110142-1G
:
Apollo Scientific
:
1-Methyl-5-nitro-1H-indazol-3-amine
APOSOR110142-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
192.175 g/mol
:
Ambient
:
73105-48-3
:
C₈H₈N₄O₂
:
MFCD16619579