2-Methyl-4-nitro-2H-indazole
Catalog # APOSOR110110-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:177.16 g/mol
Storage Temperature:Ambient
CAS Number:26120-44-5
Formula:C₈H₇N₃O₂
MDL Number:MFCD11982555