Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Italy
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
4,8-Dichloro[1]benzofuro[3,2-d]pyrimidine
4,8-Dichloro[1]benzofuro[3,2-d]pyrimidine
4,8-Dichloro[1]benzofuro[3,2-d]pyrimidine
APOSOR110089-1G
:
Apollo Scientific
:
4,8-Dichloro[1]benzofuro[3,2-d]pyrimidine
APOSOR110089-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
239.058 g/mol
:
147…149 °C
:
GHS07
:
Ambient
:
294874-71-8
:
C₁₀H₄Cl₂N₂O
:
MFCD00763947