Order Entry
Italy
ContactUsLinkComponent
2-Chloro-N-(1-methyl-1H-indazol-5-yl)acetamide
2-Chloro-N-(1-methyl-1H-indazol-5-yl)acetamide
Catalog # APOSOR110084-1G
Supplier:  Apollo Scientific
CAS Number:  
2-Chloro-N-(1-methyl-1H-indazol-5-yl)acetamide
Catalog # APOSOR110084-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :223.659 g/mol
Storage Temperature:Ambient
CAS Number:1427460-62-5
Formula:C₁₀H₁₀ClN₃O
MDL Number:MFCD22577290