Order Entry
Italy
ContactUsLinkComponent
1-Acetyl-5-(morpholin-4-ylsulphonyl)indoline
1-Acetyl-5-(morpholin-4-ylsulphonyl)indoline
Catalog # APOSOR110081-1G
Supplier:  Apollo Scientific
CAS Number:  
1-Acetyl-5-(morpholin-4-ylsulphonyl)indoline
Catalog # APOSOR110081-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :310.369 g/mol
Storage Temperature:Ambient
CAS Number:698983-77-6
Formula:C₁₄H₁₈N₂O₄S
MDL Number:MFCD05857252