Order Entry
Italy
ContactUsLinkComponent
4-[(1-Methyl-1H-indazol-3-yl)amino]-4-oxobutanoic acid
4-[(1-Methyl-1H-indazol-3-yl)amino]-4-oxobutanoic acid
Catalog # APOSOR110072-1G
Supplier:  Apollo Scientific
CAS Number:  
4-[(1-Methyl-1H-indazol-3-yl)amino]-4-oxobutanoic acid
Catalog # APOSOR110072-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :247.25 g/mol
Storage Temperature:Ambient
CAS Number:1440535-79-4
Formula:C₁₂H₁₃N₃O₃
MDL Number:MFCD24850052