Order Entry
Italy
ContactUsLinkComponent
Methyl 8-chloro-4H-thieno[3,2-c]thiochromene-2-carboxylate
Irritant
Methyl 8-chloro-4H-thieno[3,2-c]thiochromene-2-carboxylate
Catalog # APOSOR110032-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
Methyl 8-chloro-4H-thieno[3,2-c]thiochromene-2-carboxylate
Catalog # APOSOR110032-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :296.792 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:255378-11-1
Formula:C₁₃H₉ClO₂S₂
MDL Number:MFCD00106778