Order Entry
Italy
ContactUsLinkComponent
Diethyl 4-carboxyphenylphosphonate
Irritant
Diethyl 4-carboxyphenylphosphonate
Catalog # APOSOR10696-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
Diethyl 4-carboxyphenylphosphonate
Catalog # APOSOR10696-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :258.208 g/mol
Melting Pt:105…107 °C
 :GHS07
Storage Temperature:Ambient
CAS Number:1527-34-0
Formula:C₁₁H₁₅O₅P
MDL Number:MFCD01872486