Inserisci ordine
Italy
ContactUsLinkComponent
5-Bromo-2,3-dihydrobenzo[b]furan-7-sulphonyl chloride
Corrosive
5-Bromo-2,3-dihydrobenzo[b]furan-7-sulphonyl chloride
Catalog # APOSOR101632-1G
Supplier:  Apollo Scientific
CAS Number:  
Corrosive
5-Bromo-2,3-dihydrobenzo[b]furan-7-sulphonyl chloride
Catalog # APOSOR101632-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Tipo di conf.:
    Bottle
  • Conf:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :297.553 g/mol
Melting Pt:121…122 °C
Storage Temperature:Ambiente
UN:3261
ADR:8,III
 :GHS05
CAS Number:690632-00-9
Formula:C₈H₆BrClO₃S
MDL Number:MFCD04035726