
6-(Morpholin-4-yl)nicotinonitrile 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:259683-28-8
:
Formula:C₁₀H₁₁N₃O
MDL Number:MFCD00832846
:189.214 g/mol
Melting Pt:115…118 °C
Storage Temperature:Ambient
UN:3439
ADR:6.1,III
:GHS07
Frequently Bought Together