
Ethyl 5-amino-1-(1,3-benzothiazol-2-yl)-1H-pyrazole-4-carboxylate
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:186340-03-4
:
Formula:C₁₃H₁₂N₄O₂S
MDL Number:MFCD01124449
:288.325 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together