
2-[4-(1,3-Dithiolan-2-yl)phenoxy]-N'-hydroxyethanimidamide
Catalog # APOSOR018627-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:270.371 g/mol
Melting Pt:113…115 °C
:GHS07
Storage Temperature:Ambient
CAS Number:262607-85-2
Formula:C₁₁H₁₄N₂O₂S₂
MDL Number:MFCD00663347