Order Entry
Italy
ContactUsLinkComponent
1-Methylimidazo[1,5-a]pyridine
Irritant
1-Methylimidazo[1,5-a]pyridine
Catalog # APOSOR01559-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
1-Methylimidazo[1,5-a]pyridine
Catalog # APOSOR01559-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :132.163 g/mol
Boiling Pt:70 °C (3 mm Hg)
Melting Pt:64…65 °C
 :GHS07
Storage Temperature:Ambient
CAS Number:6558-62-9
Formula:C₈H₈N₂
MDL Number:MFCD06245386