
Sulpho-SASD (Sulphosuccinimidyl 2-(4-azidosalicylamido)ethyl-1,3-dithiopropionate)
Catalog # APOSBIPA102-100MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:100 mg
- Seq:2440
Specifications
About this item
:
:541.51 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:144650-95-3
Formula:C₁₆H₁₆N₅NaO₉S₃
MDL Number:MFCD01861963