
1,2:4,5-Di-O-isopropylidene-β-D-fructopyranose ≥98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:25018-67-1
:
Formula:C₁₂H₂₀O₆
MDL Number:MFCD00067639
:260.284 g/mol
Melting Pt:118…119 °C
:GHS07
Storage Temperature:Freezer
Frequently Bought Together