Order Entry
Italy
ContactUsLinkComponent
3,4,6-Tri-O-benzyl-1,2-O-(1-methoxyethylidene)-β-D-mannopyranose ≥98%
3,4,6-Tri-O-benzyl-1,2-O-(1-methoxyethylidene)-β-D-mannopyranose ≥98%
Catalog # APOSBICL4218-1G
Supplier:  Apollo Scientific
CAS Number:  
3,4,6-Tri-O-benzyl-1,2-O-(1-methoxyethylidene)-β-D-mannopyranose ≥98%
Catalog # APOSBICL4218-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :506.59 g/mol
Storage Temperature:Ambient
CAS Number:16697-49-7
Formula:C₃₀H₃₄O₇
MDL Number:MFCD00270088